CHIS5-04277

5-04277 5-04278
76229-362EA 324.26 USD
76229-362 76229-360
CHIS5-04277
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR