CHIS5-04273

5-04274 5-04273
76229-352EA 55.68 USD
76229-352 76229-354
CHIS5-04273
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR