CHIS5-04261

5-04261 5-04262
76229-328EA 204.72 USD
76229-328 76229-330
CHIS5-04261
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR