CHIS5-04237

5-04237 5-04238
76226-840EA 106.27 USD
76226-840 76226-842
CHIS5-04237
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR