CHIS5-04235

5-04235 5-04236
76226-838EA 327.54 USD
76226-838 76226-836
CHIS5-04235
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR