CHIS5-04139

5-04139 5-04140
76226-676EA 147.39 USD
76226-676 76226-678
CHIS5-04139
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR