CHIS5-04119

5-04119 5-04120
76226-620EA 180.15 USD
76226-620 76226-622
CHIS5-04119
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR