CHIS5-04111

5-04111 5-04112
76226-606EA 425.8 USD
76226-606 76226-604
CHIS5-04111
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR