CHIS5-04089

5-04089 5-04090
76226-562EA 550.27 USD
76226-562 76226-560
CHIS5-04089
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR