CHIS5-04083

5-04084 5-04083
76229-268EA 230.6 USD
76229-268 76229-270
CHIS5-04083
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR