CHIS5-04063

5-04063 5-04064
76229-228EA 98.26 USD
76229-228 76229-230
CHIS5-04063
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR