CHIS5-04055

5-04055 5-04056
76226-550EA 412.7 USD
76226-550 76226-548
CHIS5-04055
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR