CHIS5-04035

5-04036 5-04035
76226-510EA 733.69 USD
76226-510 76226-508
CHIS5-04035
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR