CHIS5-04025

5-04026 5-04025
76230-132EA 90.08 USD
76230-132 76230-134
CHIS5-04025
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR