CHIS5-04007

5-04008 5-04007
76226-100EA 573.2 USD
76226-100 76230-098
CHIS5-04007
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR