CHIS5-03997

5-03998 5-03997
76225-642EA 131.02 USD
76225-642 76222-188
CHIS5-03997
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR