CHIS5-03959

5-03959 5-03960
76225-590EA 147.39 USD
76225-590 76225-592
CHIS5-03959
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR