CHIS5-03945

5-03945 5-03946
76225-564EA 409.43 USD
76225-564 76225-562
CHIS5-03945
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR