CHIS5-03937

5-03938 5-03937
76225-546EA 139.21 USD
76225-546 76225-548
CHIS5-03937
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR