CHIS5-03919

5-03920 5-03919
76225-930EA 262.03 USD
76225-930 76225-932
CHIS5-03919
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR