CHIS5-03917

5-03917 5-03918
76225-928EA 414.33 USD
76225-928 76225-926
CHIS5-03917
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR