CHIS5-03899

5-03899 5-03900
76222-160EA 352.11 USD
76222-160 76222-158
CHIS5-03899
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR