CHIS5-03893

5-03893 5-03894
76222-148EA 786.1 USD
76222-148 76222-146
CHIS5-03893
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR