CHIS5-03865

5-03866 5-03865
76222-092EA 415.98 USD
76222-092 76222-090
CHIS5-03865
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR