CHIS5-03815

5-03815 5-03816
76222-072EA 1056.31 USD
76222-072 76222-070
CHIS5-03815
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR