CHIS5-03727

5-03727 5-03728
76225-366EA 163.77 USD
76225-366 76225-368
CHIS5-03727
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR