CHIS5-03709

5-03710 5-03709
76222-480EA 270.23 USD
76222-480 76222-478
CHIS5-03709
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR