CHIS5-03677

5-03678 5-03677
76225-292EA 270.23 USD
76225-292 76225-290
CHIS5-03677
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR