CHIS5-03667

5-03667 5-03668
76225-270EA 204.72 USD
76225-270 76225-272
CHIS5-03667
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR