CHIS5-03661

5-03661 5-03662
76225-258EA 81.89 USD
76225-258 76225-260
CHIS5-03661
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR