CHIS5-03643

5-03643 5-03644
76222-474EA 98.26 USD
76222-474 76222-476
CHIS5-03643
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR