CHIS5-03627

5-03627 5-03628
76225-224EA 491.31 USD
76225-224 76225-222
CHIS5-03627
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR