CHIS5-03589

5-03589 5-03590
76225-146EA 103.18 USD
76225-146 76225-148
CHIS5-03589
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR