CHIS5-03587

5-03588 5-03587
76225-142EA 196.52 USD
76225-142 76225-144
CHIS5-03587
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR