CHIS5-03581

5-03582 5-03581
76225-132EA 361.93 USD
76225-132 76225-130
CHIS5-03581
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR