CHIS5-03567

5-03567 5-03568
76225-102EA 65.51 USD
76225-102 76225-104
CHIS5-03567
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR