CHIS5-03563

5-03564 5-03563
76225-094EA 98.26 USD
76225-094 76225-096
CHIS5-03563
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR