CHIS5-03561

5-03561 5-03562
76225-090EA 163.77 USD
76225-090 76225-092
CHIS5-03561
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR