CHIS5-03555

5-03555 5-03556
76225-078EA 171.95 USD
76225-078 76225-080
CHIS5-03555
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR