CHIS5-03553

5-03554 5-03553
76225-076EA 319.36 USD
76225-076 76225-074
CHIS5-03553
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR