CHIS5-03535

5-03535 5-03536
76225-038EA 57.32 USD
76225-038 76225-040
CHIS5-03535
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR