CHIS5-03525

5-03525 5-03526
76225-020EA 491.31 USD
76225-020 76225-018
CHIS5-03525
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR