CHIS5-03511

5-03512 5-03511
76224-992EA 691.11 USD
76224-992 76224-990
CHIS5-03511
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR