CHIS5-03497

5-03498 5-03497
76224-964EA 409.43 USD
76224-964 76224-962
CHIS5-03497
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR