CHIS5-03463

5-03463 5-03464
76224-896EA 417.62 USD
76224-896 76224-894
CHIS5-03463
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR