CHIS5-03457

5-03457 5-03458
76222-440EA 224.36 USD
76222-440 76222-438
CHIS5-03457
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR