CHIS5-03417

5-03418 5-03417
76224-810EA 425.8 USD
76224-810 76224-812
CHIS5-03417
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR