CHIS5-03403

5-03404 5-03403
76224-784EA 818.85 USD
76224-784 76224-782
CHIS5-03403
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR