CHIS5-03383

5-03384 5-03383
76224-752EA 589.57 USD
76224-752 76224-750
CHIS5-03383
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR