CHIS5-03377

5-03377 5-03378
76224-740EA 736.97 USD
76224-740 76224-738
CHIS5-03377
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR